Learn More
Phenylbiguanide, MedChemExpress
Product Code. 16404586
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
16404586 100 mg 100mg
1 options
Product Code. 16404586 Supplier Medchem Express Supplier No. HY101331
 Restricted Item
Beräknad leverans: 30-11-2026
MedChemExpress Phenylbiguanide is a 5-HT3 receptor selective agonist with an EC50 of 3.0±0.1 μM.
Phenylbiguanide is a 5-HT3 receptor selective agonist with an EC50 of 3.0±0.1 μM.
Specifications
Chemical Name or Material Phenylbiguanide
Physical Form Solid
CAS 102-02-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H11N5
For Use With (Application) Neuroscience-Neuromodulation
Synonym N-Phenylbiguanide PBG 1-Phenylbiguanide
Solubility Information DMSO : 150 mg/mL (846.45 mM; Need ultrasonic)
SMILES NC(NC(NC1=CC=CC=C1)=N)=N
Molecular Weight (g/mol) 177.21
Formula Weight 177.21
Percent Purity 98.0%
Grade Research