Läs mer
Phenylbutazone EP Impurity A, Mikromol™
Produktkod. 30050208
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050208 100 mg
30051154 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30050208 Leverantör Mikromol Leverantörsnummer MM0548.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Phenylbutazone EP Impurity A
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2RS)-2-[(1,2-Diphenyldiazanyl)carbonyl]hexanoic Acid
CAS 3583-64-0
Analyt- eller komponentnamn (2RS)-2-[(1,2-Diphenyldiazanyl)carbonyl]hexanoic Acid
Frakt skick Ice pack (-20°C)
Molekylformel C19 H22 N2 O3
Certifikat och efterlevnad ISO 17025
Synonym (2RS)-2-[(1,2-Diphenyldiazanyl)carbonyl]hexanoic Acid, Phenylbutazone Imp. A (EP), Propanedioic acid, 2-butyl-, 1-(1,2-diphenylhydrazide), Malonic acid, butyl-, 1,2-diphenylhydrazide (7CI), Malonic acid, butyl-, mono(1,2-diphenylhydrazide) (8CI), Propanedioic acid, butyl-, mono(1,2-diphenylhydrazide) (9CI), Bumadizon, Bumadizone, Butylmalonic acid mono(1,2-diphenylhydrazide), N-(2-Carboxycaproyl)hydrazobenzene, Phenylbutazone Impurity A
InChI formel InChI=1S/C19H22N2O3/c1-2-3-14-17(19(23)24)18(22)21(16-12-8-5-9-13-16)20-15-10-6-4-7-11-15/h4-13,17,20H,2-3,14H2,1H3,(H,23,24)
LEDER CCCCC(C(=O)O)C(=O)N(Nc1ccccc1)c2ccccc2
IUPAC-namn 2-[anilino(phenyl)carbamoyl]hexanoic acid
Molekylvikt (g/mol) 326.39
Kvantitet 100 mg
Formel vikt 326.163 g/mol