Läs mer
Phenylmethyl Ester 2-[1-(3,4-Dimethoxybenzoyl)-2-(1-ethylpropoxy)-1-methyl-2-oxoethyl] Hydrazinecarboxylic Acid, TRC
Produktkod. 30415125
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30415125 25 mg
1 options
Produktkod. 30415125 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP43186525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phenylmethyl Ester 2-[1-(3,4-Dimethoxybenzoyl)-2-(1-ethylpropoxy)-1-methyl-2-oxoethyl] Hydrazinecarboxylic Acid
CAS 1281895-71-3
Molekylformel C25H32N2O7
Kvantitet 25 mg
Synonym 2-[1-(3,4-Dimethoxybenzoyl)-2-(1-ethylpropoxy)-1-methyl-2-oxoethyl]-phenylmethyl Ester Hydrazinecarboxylic acidPhenylmethyl 2-[1-(3,4-Dimethoxybenzoyl)-2-(1-ethylpropoxy)-1-methyl-2-oxoethyl]hydrazinecarboxylate
InChI formel InChI=1S/C25H32N2O7/c1-6-19(7-2)34-23(29)25(3,22(28)18-13-14-20(31-4)21(15-18)32-5)27-26-24(30)33-16-17-11-9-8-10-12-17/h8-15,19,27H,6-7,16H2,1-5H3,(H,26,30)/t25-/m1/s1
LEDER CCC(CC)OC(=O)[C@](C)(NNC(=O)OCc1ccccc1)C(=O)c2ccc(OC)c(OC)c2
IUPAC-namn pentan-3-yl (2R)-3-(3,4-dimethoxyphenyl)-2-methyl-3-oxo-2-(2-phenylmethoxycarbonylhydrazinyl)propanoate
Molekylvikt (g/mol) 472.54
Formel vikt 472.221