Läs mer
Phenylmethyl Ester [4-(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)butyl][3-[[(phenylmethoxy)carbonyl]amino]propyl]carbamic Acid, TRC
Produktkod. 30371924
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30371924 5 mg
1 options
Produktkod. 30371924 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD9762805MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phenylmethyl Ester (4-(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)butyl)(3-(((phenylmethoxy)carbonyl)amino)propyl)carbamic Acid
CAS 97628-83-6
Molekylformel C31H33N3O6
Kvantitet 5 mg
Synonym Benzyl (3-(((Benzyloxy)carbonyl)amino)propyl)(4-(1,3-dioxoisoindolin-2-yl)butyl)carbamate
InChI formel InChI=1S/C31H33N3O6/c35-28-26-16-7-8-17-27(26)29(36)34(28)21-10-9-19-33(31(38)40-23-25-14-5-2-6-15-25)20-11-18-32-30(37)39-22-24-12-3-1-4-13-24/h1-8,12-17H,9-11,18-23H2,(H,32,37)
LEDER O=C(NCCCN(CCCCN1C(=O)c2ccccc2C1=O)C(=O)OCc3ccccc3)OCc4ccccc4
IUPAC-namn benzyl N-[4-(1,3-dioxoisoindol-2-yl)butyl]-N-[3-(phenylmethoxycarbonylamino)propyl]carbamate
Molekylvikt (g/mol) 543.61
Formel vikt 543.2369