missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylselenyl chloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
10 g
25 g
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Melting Point | 61.0°C to 65.0°C |
Color | Orange |
Boiling Point | 120.0°C (20.0 mmHg) |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (Argentometry) |
Packaging | Glass bottle |
Linear Formula | C6H5SeCl |
Quantity | 1 g |
Beilstein | 06, III, 1110 |
Fieser | 05,518; 06,459; 07,286; 09,25; 11,33; 13,26; 16,19; 17,26 |
Afficher plus |
Sécurité et traitement
Identifiant produit
- Phenylselenyl chloride
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H373-May cause damage to organs.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'