Läs mer
(Phenylthio)methyltriphenylphosphonium Chloride, TRC
Produktkod. 30426856
Change view
Kvantitet:
2 g
Produktkod. Kvantitet
30426856 2 g
1 options
Produktkod. 30426856 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3370002G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (Phenylthio)methyltriphenylphosphonium Chloride
CAS 13884-92-9
Molekylformel C25 H22 P S . Cl
Kvantitet 2 g
InChI formel InChI=1S/C25H22PS.ClH/c1-5-13-22(14-6-1)26(23-15-7-2-8-16-23,24-17-9-3-10-18-24)21-27-25-19-11-4-12-20-25;/h1-20H,21H2;1H/q+1;/p-1
LEDER [Cl-].C(Sc1ccccc1)[P+](c2ccccc2)(c3ccccc3)c4ccccc4
IUPAC-namn triphenyl(phenylsulfanylmethyl)phosphanium;chloride
Molekylvikt (g/mol) 420.93
Formel vikt 420.0868