Läs mer
Phenyltoloxamine Citrate Salt, TRC
Produktkod. 30421013
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30421013 50 mg
1 options
Produktkod. 30421013 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP33737050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phenyltoloxamine Citrate
CAS 1176-08-5
Molekylformel C17 H21 N O . C6 H8 O7
Kvantitet 50 mg
Synonym Phenyltoloxamine Citrate
InChI formel InChI=1S/C17H21NO.C6H8O7/c1-18(2)12-13-19-17-11-7-6-10-16(17)14-15-8-4-3-5-9-15;7-3(8)1-6(13,5(11)12)2-4(9)10/h3-11H,12-14H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)
LEDER CN(C)CCOc1ccccc1Cc2ccccc2.OC(=O)CC(O)(CC(=O)O)C(=O)O
IUPAC-namn 2-(2-benzylphenoxy)-N,N-dimethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
Molekylvikt (g/mol) 447.48
Formel vikt 447.1893