Learn More
Phenyltrimethylammonium iodide, 99%, Thermo Scientific Chemicals
Catalog Number p-7021881
Click to view available options
Quantity:
25 g
100 g
500 g
98-04-4
C9H14IN
263.122
MFCD00011791
KKLAORVGAKUOPZ-UHFFFAOYSA-M
phenyltrimethylammonium iodide, trimethylphenylammonium iodide, n,n,n-trimethylanilinium iodide, trimethylanilinium iodide, trimethylanillinium iodide, n,n-dimethylaniline methiodide, unii-4o888xq7fp, ammonium, phenyltrimethyl-, iodide, n,n,n-trimethylbenzenaminium iodide, benzenaminium, n,n,n-trimethyl-, iodide
94135
trimethyl(phenyl)azanium;iodide
C[N+](C)(C)C1=CC=CC=C1.[I-]
98-04-4
C9H14IN
263.122
MFCD00011791
KKLAORVGAKUOPZ-UHFFFAOYSA-M
phenyltrimethylammonium iodide, trimethylphenylammonium iodide, n,n,n-trimethylanilinium iodide, trimethylanilinium iodide, trimethylanillinium iodide, n,n-dimethylaniline methiodide, unii-4o888xq7fp, ammonium, phenyltrimethyl-, iodide, n,n,n-trimethylbenzenaminium iodide, benzenaminium, n,n,n-trimethyl-, iodide
94135
trimethyl(phenyl)azanium;iodide
C[N+](C)(C)C1=CC=CC=C1.[I-]
Phenyltrimethylammonium iodide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenyltrimethylammonium iodide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.
Solubility
Soluble in water and methanol.
Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. It is hygroscopic in nature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∼227°C (sublimation)
Quantity 25 g
UN Number UN2811
Beilstein 3916752
Merck Index 14,7318
Formula Weight 263.12
Percent Purity 99%
Chemical Name or Material Phenyltrimethylammonium iodide