Learn More
Phenyramidol Hydrochloride, LoGiCal
Product Code. 30457473
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
30457473 10 mg 10mg
1 options
Product Code. 30457473 Supplier LoGiCal Supplier No. LGCFOR3188.00
 Restricted Item
Beräknad leverans: 30-11-2026
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Phenyramidol Hydrochloride
CAS 326-43-2
Analyte or Component Names Phenyramidol Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C13 H14 N2 O . Cl H
Certifications/Compliance ISO 9001
Synonym Abbolexin, Anabloc, Analexin, Betapirin, Elan, Fenyramidol hydrochloride, Firmalgil, IN 511, MJ 505, Miodar, NSC 17777, Phenylramidol hydrochloride, Phenyramidol hydrochloride, Vazalmin, Verbanol, Benzenemethanol, alpha-[(2-pyridinylamino)methyl]-, hydrochloride (1:1), Benzenemethanol, alpha-[(2-pyridinylamino)methyl]-, monohydrochloride (9CI), Benzyl alcohol, alpha-[(2-pyridylamino)methyl]-, hydrochloride (7CI), Benzyl alcohol, alpha-[(2-pyridylamino)methyl]-, monohydrochloride (6CI,8CI), alpha-[(2-Pyridylamino)methyl]benzyl alcohol hydrochloride
InChI Formula InChI=1S/C13H14N2O.ClH/c16-12(11-6-2-1-3-7-11)10-15-13-8-4-5-9-14-13;/h1-9,12,16H,10H2,(H,14,15);1H
Solution Type Neat
SMILES Cl.OC(CNc1ccccn1)c2ccccc2
IUPAC Name 1-phenyl-2-(pyridin-2-ylamino)ethanol;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 250.72
Quantity 10 mg
Formula Weight 250.0873 g/mol