Läs mer
Pheophorbide A (>80%) (mixture of diastereomers), TRC
Produktkod. 30134143
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134143 25 mg
30007984 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134143 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP99842825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 15664-29-6
Molekylformel C35 H36 N4 O5
Molekylvikt (g/mol) 592.68
Synonym 2-Deacetyl-2-vinylbacteriopheophorbide, Phaeophorbide A, Pheophorbide a5
LEDER CCc1c(C)c2cc3[nH]c(cc4nc([C@@H](CCC(=O)O)[C@@H]4C)c5[C@@H](C(=O)OC)C(=O)C6=C(C)c(cc1n2)[nH]c56)c(C)c3C=C
Specifikationer
Kemiskt namn eller material Pheophorbide A
Kvantitet 25 mg
InChI formel InChI=1S/C35H36N4O5/c1-8-19-15(3)22-12-24-17(5)21(10-11-28(40)41)32(38-24)30-31(35(43)44-7)34(42)29-18(6)25(39-33(29)30)14-27-20(9-2)16(4)23(37-27)13-26(19)36-22/h8,12-14,17,21,31,36,39H,1,9-11H2,2-7H3,(H,40,41)/b22-12-,23-13-,24-12-,25-14-,26-13-,27-14-,32-30-/t17-,21-,31+/m0/s1
Formel vikt 592.2686