Beskrivning
- Sporty safety spectacles with high wearer acceptance
- Robust polycarbonate lens with proven uvex supra-vision coating technology - permanently anti-fog and scratch-resistant
- Metal-free
- Comfort features X-tended eyeshield: Soft-seal frame technology wraps around the forehead and provides protection against particle penetration
- Ergonomically shaped side arms with soft ear-pieces ensure a comfortable and secure fit without creating pressure points
- Soft, flexible nose piece adapts to the wearer to deliver a non-slip, pressure-free fit
- Certified in accordance with EN 166 (personal eye protection) and EN 170 (UV filters) UV400 -100% protection against harmful UV radiation up to 400 nm
Specifikationer
Specifikationer
| Linsbeläggning | Anti-fog, Anti-scratch, Scratch-resistant |
| Linsfärg | Clear |
| Produkttyp | Safety Glasses |
| Material | Polycarbonate |
| Disponibel | Reusable |
| Linsmaterial | Polycarbonate |
| Rammaterial | TPE |
| UV-skydd | UV400 |
| Ramtyp | Semi-rimless |