Description
The attractive design is underpinned by innovative safety eyewear technology: The main feature of the spectacles is the duo-spherical lens.This lens not only offers a wide field of vision, but it is also particularly scratch-resistant on the outside and features permanent anti-fog protection on the inside thanks to high-quality uvex supra-vision coating technology. By adding an extra frame and headband, the uvex pheos can be expanded into a set, depending on the field of use.
- Sporty, timeless safety spectacles
- Frameless design affords an extra-large field of vision
- Robust polycarbonate lens with proven uvex supra-vision coating technology - permanently anti-fog and scratch-resistant
- Optional extra frame offers additional protection against particles, dirt and dust
- Optional headband keeps the spectacles in position during demanding work
- Narrow version, also available in standard size
- Fits uvex IES Metal-free
- Ergonomically shaped side arms with soft ear-pieces ensure a comfortable, secure fit without creating pressure points
- Anatomically shaped nose piece for all-day wearer comfort
- Certified in accordance with EN 166 (personal eye protection) and EN 172 (sunglare filters for industrial use) UV400 - 100% protection against harmful UV radiation to 400 nm
Specifications
Specifications
| Linsbeläggning | Anti-fog, Scratch-resistant, Anti-scratch |
| Linsfärg | Gray |
| Produkttyp | Safety Glasses |
| Material | Polycarbonate |
| Disponibel | Reusable |
| Linsmaterial | Polycarbonate |
| Rammaterial | TPE |
| UV-skydd | UV400 |
| Ramtyp | Semi-rimless |