Learn More
uvex Pheos Visor with Chin Guard
Product Code. 17777739 Shop All Uvex Products
Change view
Click to view available options
Color: Clear
Product Code.
17777739 Color: Clear
1 options
Product Code. 17777739 Supplier uvex Supplier No. 9906009
  • ONLINE EXCLUSIVE PRICE
    1330.00 SEK
    1257.36 SEK / Paket med 1
    Save 72.64 SEK
    5% Av
Beräknad leverans: 27-11-2026
Lightweight, robust visor for uvex pheos safety helmets B-S-WR and uvex pheos alpine.
  • For pheos alpine a visior adapter 9790 067 is required
  • Automatic magnetic lock, flip-up mechanism with park position, lens replacement
  • Lens PC clear/UV 400, 2C-1,2 W1 B 9 KN CE
  • Coating uvex supravision excellence, outside anti-scratch, inside anti-fog
  • Helmet not included
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Polycarbonate
Lens Coating Supravision Excellence