Läs mer
Invitrogen™ PHF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597060
Beskrivning
Immunogen sequence: MAQPPRLSRS GASSLWDPAS PAPTSGPRPR LWEGQDVLAR WTDGLLYLGT IKKVDSAREV CLVQFEDDSQ FLVLWKDISP AALPGEELLC CVCRSETVVP GNRLVSCEKC RHAYHQDCHV PRAPAPGEGE GTSWVCRQCV FAIATKRGGA LKKGPYARAM LGMKLSLPYG LKGLDWDAGH LSNRQQSYCY CGGPGEWNLK MLQCRSCLQW FHEACTQCLS KPLLYGDRFY EFECCVCRGG PEKVRRLQLR WVDVAHLVLY; Positive Samples: A-549, HeLa, Mouse lung; Cellular Location: Cytoplasm, Nucleus, centrosome, cytoskeleton, microtubule organizing center.
This gene encodes a protein with significant sequence similarity to Drosophila Polycomblike. The encoded protein contains a zinc finger-like PHD (plant homeodomain) finger which is distinct from other classes of zinc finger motifs and which shows the typical Cys4-His-Cys3 arrangement. PHD finger genes are thought to belong to a diverse group of transcriptional regulators possibly affecting eukaryotic gene expression by influencing chromatin structure. Two transcript variants have been found for this gene.
Specifikationer
| PHF1 | |
| Polyclonal | |
| Unconjugated | |
| PHF1 | |
| AW557215; D17Ertd455e; hPCl1; hPHF1; mPCl1; MTF2L2; PCL1; PHD finger protein 1; PHF1; Phf2; Plc1; polycomb-like 1; polycomb-like protein 1; Protein PHF1; T-complex testis-expressed 3; Tctex3; Tctex-3; TDRD19C; testicular tissue protein Li 140; tudor domain containing 19C | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 21652, 294287, 5252 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.87 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43189, Q9Z1B8 | |
| PHF1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human PHF1 (NP_0770841). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.