Läs mer
Phillyrin, TRC
Produktkod. 30423485
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30423485 100 mg
1 options
Produktkod. 30423485 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP338400100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phillyrin
CAS 487-41-2
Molekylformel C27 H34 O11
Kvantitet 100 mg
Synonym β-D-Glucopyranoside','4-[(1S,3aR,4R,6aR)-4-(3,4-dimethoxyphenyl)tetrahydro-1H,3H-furo[3,4-c]furan-1-yl]-2-methoxyphenyl, Phillyrin (8CI), Phyllyrin (6CI), β-D-Glucopyranoside','4-[4-(3,4-dimethoxyphenyl)tetrahydro-1H,3H-furo[3,4-c]furan-1-yl]-2-methoxyphenyl','[1S-(1α,3aα,4β,6aα)]-, 1H,3H-Furo[3,4-c]furan','β-D-glucopyranoside deriv., 4-[(1S,3aR,4R,6aR)-4-(3,4-Dimethoxyphenyl)tetrahydro-1H,3H-furo[3,4-c]furan-1-yl]-2-methoxyphenyl β-D-glucopyranoside, Phillygenin β-D-glucopyranoside, Phillyroside
InChI formel InChI=1S/C27H34O11/c1-32-17-6-4-13(8-19(17)33-2)25-15-11-36-26(16(15)12-35-25)14-5-7-18(20(9-14)34-3)37-27-24(31)23(30)22(29)21(10-28)38-27/h4-9,15-16,21-31H,10-12H2,1-3H3/t15-,16-,21+,22+,23-,24+,25-,26+,27+/m0/s1
LEDER COc1ccc(cc1OC)[C@@H]2OC[C@H]3[C@@H]2CO[C@@H]3c4ccc(O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)c(OC)c4
IUPAC-namn (2S,3R,4S,5S,6R)-2-[4-[(3R,3aR,6S,6aR)-3-(3,4-dimethoxyphenyl)-1,3,3a,4,6,6a-hexahydrofuro[3,4-c]furan-6-yl]-2-methoxyphenoxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Molekylvikt (g/mol) 534.55
Formel vikt 534.2101