missing translation for 'onlineSavingsMsg'
Läs mer

Phloroglucin Tribenzyl Ether, TRC

Produktkod. 30419196
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
2.5 mg
Förpackningsstorlek:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30419196 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30419196 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3400202.5MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Phloroglucin Tribenzyl Ether
CAS 59434-20-7
Molekylformel C27H24O3
Kvantitet 2.5 mg
Synonym 1,3,5-Tris(phenylmethoxy)benzene, 1,3,5-Tribenzyloxybenzene, 1,3,5-Tris(benzyloxy)benzene, Tribenzyl Phloroglucinol
InChI formel InChI=1S/C27H24O3/c1-4-10-22(11-5-1)19-28-25-16-26(29-20-23-12-6-2-7-13-23)18-27(17-25)30-21-24-14-8-3-9-15-24/h1-18H,19-21H2
LEDER C(Oc1cc(OCc2ccccc2)cc(OCc3ccccc3)c1)c4ccccc4
IUPAC-namn 1,3,5-tris(phenylmethoxy)benzene
Molekylvikt (g/mol) 396.48
Formel vikt 396.1725
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.