Läs mer
Phloroglucin Tribenzyl Ether, TRC
Produktkod. 30417160
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30417160 10 mg
1 options
Produktkod. 30417160 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP34002010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phloroglucin Tribenzyl Ether
CAS 59434-20-7
Molekylformel C27H24O3
Kvantitet 10 mg
Synonym 1,3,5-Tris(phenylmethoxy)benzene, 1,3,5-Tribenzyloxybenzene, 1,3,5-Tris(benzyloxy)benzene, Tribenzyl Phloroglucinol
InChI formel InChI=1S/C27H24O3/c1-4-10-22(11-5-1)19-28-25-16-26(29-20-23-12-6-2-7-13-23)18-27(17-25)30-21-24-14-8-3-9-15-24/h1-18H,19-21H2
LEDER C(Oc1cc(OCc2ccccc2)cc(OCc3ccccc3)c1)c4ccccc4
IUPAC-namn 1,3,5-tris(phenylmethoxy)benzene
Molekylvikt (g/mol) 396.48
Formel vikt 396.1725