Läs mer
Phloroglucinol-13C6, TRC
Produktkod. 30422186
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30422186 10 mg
30409684 1 mg
30410088 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30422186 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP34000210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1329497-24-6
Molekylformel 13C6H6O3
Molekylvikt (g/mol) 132.07
Synonym 1,3,5-Benzenetriol-13C6, 1,3,5-Trihydroxybenzene-13C6, Benzene-s-triol-13C6, Dilospan S-13C6, NSC 1572-13C6, Phloroglucin-13C6, Phloroglucine-13C6, Spasfon-Lyoc-13C6, sym-Trihydroxybenzene-13C6
IUPAC-namn (1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-triene-1,3,5-triol
LEDER O[13c]1[13cH][13c](O)[13cH][13c](O)[13cH]1
Specifikationer
Kemiskt namn eller material Phloroglucinol-13C6
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C6H6O3/c7-4-1-5(8)3-6(9)2-4/h1-3,7-9H/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vikt 132.0518
Procent renhet >95%