Läs mer
Phosfolan-methyl 100 μg/mL in Methanol, Dr. Ehrenstorfer
Specifikationer
Specifikationer
| Kemiskt namn eller material | Phospholan-methyl |
| CAS | 5120-23-0 |
| Analyt- eller komponentnamn | Phospholan-methyl |
| Frakt skick | +20°C |
| Molekylformel | C5 H10 N O3 P S2 |
| Koncentration | 100 μg/mL |
| Certifikat och efterlevnad | ISO 17034 |
| Synonym | Imidocarbonic acid, phosphonodithio-, cyclic ethylene P,P-dimethyl ester (7CI,8CI), Phosphoramidic acid, 1,3-dithiolan-2-ylidene-, dimethyl ester (9CI), 1,2-Ethanedithiol, cyclic ester with phosphonodithioimidocarbonic acid P,P-dimethyl ester, Phosfolan-methyl, Phosphoramidic acid, N-1,3-dithiolan-2-ylidene-, dimethyl ester |
| InChI formel | InChI=1S/C5H10NO3PS2/c1-8-10(7,9-2)6-5-11-3-4-12-5/h3-4H2,1-2H3 |
| LEDER | COP(=O)(OC)N=C1SCCS1 |
| Visa mer |