Läs mer
Phosfolan, TRC
Produktkod. 30425574
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425574 250 mg
30424630 50 mg
30428099 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30425574 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP365003250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 947-02-4
Molekylformel C7 H14 N O3 P S2
Molekylvikt (g/mol) 255.29
Synonym Imidocarbonic acid, phosphonodithio-, cyclic ethylene P,P-diethyl ester (7CI,8CI), Phosphoramidic acid, 1,3-dithiolan-2-ylidene-, diethyl ester (9CI), (Diethoxyphosphinyl)dithioimidocarbonic acid cyclic ethylene ester, 2-[Diethoxyphosphinylimino]-1,3-dithiolane, AC 47031, American Cyanamide 47031, CL 47031, Cyolane, Diethyl 1,3-dithiolan-2-ylidenephosphoramidate, EI 47031, ENT 25,830, Phosfolan, Phospholan
IUPAC-namn N-diethoxyphosphoryl-1,3-dithiolan-2-imine
LEDER CCOP(=O)(OCC)N=C1SCCS1
Specifikationer
Kemiskt namn eller material Phosfolan
Kvantitet 250 mg
InChI formel InChI=1S/C7H14NO3PS2/c1-3-10-12(9,11-4-2)8-7-13-5-6-14-7/h3-6H2,1-2H3
Formel vikt 255.0153