missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Phospho-4EBP1 (Thr37, Thr46) Polyclonal Antibody
GREENER_CHOICE

Produktkod. 11582620
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
Förpackningsstorlek:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11582620 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11582620 Leverantör Invitrogen™ Leverantörsnummer PA517728

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

This gene encodes one member of a family of translation repressor proteins. The protein directly interacts with eukaryotic translation initiation factor 4E, which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5' end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Phospho-4EBP1 (Thr37, Thr46)
Användningsområden Western Blot
Klassificering Polyclonal
Koncentration 197 μg/mL
Konjugera Unconjugated
Formulering 0.01M HEPES with 0.15M NaCl, 100 μg/mL BSA, 50% glycerol and no preservative; pH 7.5
Gen EIF4EBP1
Genaccessionsnr. Q13541, Q60876, Q62622
Gene Alias 4EBP1; 4E-BP1; AA959816; BP-1; eIF4E-binding protein 1; EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1; eukaryotic translation initiation factor 4E-binding protein 1; MGC431; MGC4316; P/OKCL.6; PHAS-1; PHAS-I; Phosphorylated heat- and acid-stable protein regulated by insulin 1
Gensymboler EIF4EBP1
Värd art Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding pThr37 of mouse 4E-BP1 and pThr46 of mouse 4E-BP1.
Reningsmetod Antigen affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 116636, 13685, 1978
Målarter Human, Mouse, Monkey, Rat
Innehåll och lagring -20°C
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.