Learn More
Phospho-BTK/ITK (Tyr551, Tyr511) Monoclonal Antibody (M4G3LN), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 12-9015-42
Additional Details : Weight : 0.23750kg
Description
Description: This M4G3LN monoclonal antibody recognizes human and mouse Bruton's tyrosine kinase (also known as BTK, agammaglobulinaemia tyrosine kinase, B-cell progenitor kinase)/interleukin-2-inducible tyrosine kinase (also known as ITK, T-cell-specific kinase, Tyrosine-protein kinase Lyk) when phosphorylated on Y551 and Y511, respectively. In addition to Src family and SYK family kinases, B cells and T cells express a third family of receptor-proximal kinases called the Tec kinase family. The B cell-specific Tec family kinase is BTK; T cells express ITK as well as Tec and Rlk. Tec family kinases play an indispensable role in activation of phospholipase C and activation of calcium-dependent signaling cascades following antigen receptor triggering. BTK has been implicated in propagating signals downstream of LPS-TLR4 in B cells via signaling through the TLR4 co-receptor RP105/CD180. Mice congenitally lacking BTK or ITK show profound defects in B cell or T cell development and signaling, respectively. People with mutations or deletions in BTK suffer from X-linked agammaglobulinemia and X-linked immunodeficiency. Specificity of this M4G3LN clone was determined by ELISA, flow cytometry, and western blotting. Applications Reported: This M4G3LN antibody has been reported for use in intracellular staining followed by flow cytometric analysis.
In addition to Src family and SYK family kinases, B cells and T cells express a third family of receptor-proximal kinases called the Tec kinase family. The B cell-specific Tec family kinase is BTK; T cells express ITK as well as Tec and Rlk. Tec family kinases play an indispensable role in activation of phospholipase C and activation of calcium-dependent signaling cascades following antigen receptor triggering. BTK has been implicated in propagating signals downstream of LPS-TLR4 in B cells via signaling through the TLR4 co-receptor RP105/CD180. Mice congenitally lacking BTK or ITK show profound defects in B cell or T cell development and signaling, respectively. People with mutations or deletions in BTK suffer from X-linked agammaglobulinemia and X-linked immunodeficiency.Specifications
Phospho-BTK/ITK (Tyr551, Tyr511) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P35991, Q03526, Q06187, Q08881 | |
BTK, ITK | |
Affinity chromatography | |
RUO | |
12229, 16428, 3702, 695 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
M4G3LN | |
PE | |
BTK | |
agammaglobulinaemia tyrosine kinase; agammaglobulinemia tyrosine kinase; AGMX1; AI528679; AT; ATK; B-cell progenitor kinase; BPK; Bruton agammaglobulinemia tyrosine kinase; bruton tyrosine kinase; Bruton's tyrosine kinase; BTK; dominant-negative kinase-deficient Brutons tyrosine kinase; Emt; homolog of mouse T-cell itk/tsk; IL2 inducible T cell kinase; IL2 inducible T-cell kinase; IL-2-inducible T cell kinase; IL-2-inducible T-cell kinase; IL2-inducible T-cell kinase; IMD1; interleukin-2-inducible T cell kinase; interleukin-2-inducible T-cell kinase; ITK; Kinase EMB; kinase EMT; Kinase TLK; LPFS1; LYK; PSCTK1; PSCTK2; T-cell-specific kinase; Tcsk; Tlk; truncated Bruton agammaglobulinemia tyrosine kinase; Tsk; Tyrosine-protein kinase BTK; Tyrosine-protein kinase BTK (Brutons tyrosine kinase) (Agammaglobulinaemia tyrosine kinase) (ATK) (B cell progenitor kinase) (BPK) (Kinase EMB); tyrosine-protein kinase BTK isoform (lacking exon 13 to 17); tyrosine-protein kinase BTK isoform (lacking e | |
Mouse | |
100 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b κ |
For Research Use Only.