missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Cardiac Troponin I (Ser22, Ser23) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540251
Additional Details : Weight : 0.00910kg
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
Phospho-Cardiac Troponin I (Ser22, Ser23) Polyclonal antibody specifically detects Phospho-Cardiac Troponin I (Ser22, Ser23) in Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
Phospho-Cardiac Troponin I (Ser22, Ser23) | |
Polyclonal | |
Unconjugated | |
TNNI3 | |
alternatively spliced; cardiac ankyrin repeat kinase; Cardiac muscle troponin T; cardiac troponin I; cardiac troponin I isoform; cardiac troponin I-interacting kinase; cardiac troponin T; cardiac troponin T isoform 1; cardiac troponin T isoform 2; cardiac troponin T2; cardiomyopathy, dilated 1D (autosomal dominant); cardiomyopathy, dilated 2A (autosomal recessive); cardiomyopathy, hypertrophic 2; CARK; CCDD; CMD1D; CMD1FF; CMD2A; CMH2; CMH7; CMPD2; cTnI; cTnT; Ctt; CTTG; D830019J24Rik; LVNC6; MGC116817; RATCTTG; RCM1; RCM3; Serine/threonine-protein kinase TNNI3K; Tn1; TnI; TNNC1; Tnni3; TNNI3 interacting kinase; TNNI3-interacting kinase; TNNI3K; tnni3k {ECO:0000250; TNNT2; Tnnt3; TnTc; Tpn1; troponin 1, type 3; troponin I type 3 (cardiac); troponin I, cardiac 3; troponin I, cardiac muscle; troponin I, cardiac muscle-like; troponin I3, cardiac type; Troponin T cardiac; troponin T cardiac isoform; troponin T isoform 1; troponin T isoform 2; troponin T splicing mutant isoform 1; troponin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21954, 29248 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P23693, P48787 | |
TNNI3 | |
Synthesized peptide derived from human Troponin I-C around the phosphorylation site of S22/S23. | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |