missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Cyclin B1 (Ser128) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564671
Additional Details : Weight : 0.01000kg
Description
Phospho-Cyclin B1 (Ser128) Polyclonal Antibody detects endogenous levels of Cyclin B1 only when phosphorylated at Ser128.
Phospho-Cyclin B1 (Ser128) Polyclonal antibody specifically detects Phospho-Cyclin B1 (Ser128) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Phospho-Cyclin B1 (Ser128) | |
Polyclonal | |
Unconjugated | |
Ccnb1 | |
Ccn-2; CCNB; Ccnb1; Ccnb1-rs1; Ccnb1-rs13; Cycb; Cycb1; Cycb1-rs1; Cycb-4; Cycb-5; cyclin B; cyclin B1; cyclin B1, related sequence 1; cyclin B1, related sequence 13; G2/mitotic-specific cyclin B1; G2/mitotic-specific cyclin-B1; I79_013589; OTTHUMP00000221981; OTTHUMP00000223023 | |
Rabbit | |
Affinity chromatography | |
RUO | |
25203, 268697, 891 | |
-20°C | |
Liquid |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 30% glycerol and 0.02% sodium azide; pH 7.3 | |
P14635, P24860, P30277 | |
Ccnb1 | |
A synthesized peptide derived from human Cyclin B1 (Phospho-Ser128) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |