missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PERK (Thr982) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540294
Additional Details : Weight : 0.00910kg
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
Phospho-PERK (Thr982) Polyclonal antibody specifically detects Phospho-PERK (Thr982) in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
Phospho-PERK (Thr982) | |
Polyclonal | |
Unconjugated | |
EIF2AK3 | |
2.7.11.1; AI427929; EIF2AK3; eukaryotic translation initiation factor 2 alpha kinase 3; eukaryotic translation initiation factor 2-alpha kinase 3; HsPEK; OTTHUMP00000207187; OTTHUMP00000207188; Pancreatic eIF2-alpha kinase; PEK; Perk; PRKR-like endoplasmic reticulum kinase; WRS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13666, 29702, 9451 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9NZJ5, Q9Z1Z1, Q9Z2B5 | |
EIF2AK3 | |
Synthesized peptide derived from human PERK around the phosphorylation site of T982. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |