missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Phospho-Histone H1 (Thr3) Polyclonal Antibody

Beskrivning
This antibody is predicted to react with mouse and rat based on sequence homology.
This locus is the ortholog of a mouse protein-coding gene that displays characteristics of a linker histone and is expressed in nuclei of late maturing spermatids. The human locus is expressed; however, the open reading frame has been disrupted by a frameshift so it is unlikely to encode a functional protein.
Specifikationer
Specifikationer
| Antigen | Phospho-Histone H1 (Thr3) |
| Användningsområden | Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugera | Unconjugated |
| Formulering | Whole serum with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gen | H1-5 |
| Genaccessionsnr. | D3ZBN0, D4A3K5, P07305, P10922, P15864, P16401, P16403, P43275, P43276, P43278, Q02539 |
| Gene Alias | 0610008C09Rik; CTL-1; H1; H1 antibody; H1 histone family member 0; H1 histone family, member 0; H1 histone family, member 1; H1 histone family, member 2; H1 histone family, member 5; H1 VAR.1; H1 VAR.3; H1 VAR.5; H1(0); H1.0; H1.0, H1(0), H1-0; H1.1; H1.12 linker histone, cluster member; H1.2; H1.2 linker histone, cluster member; H1.5; H1.9; H10; H1-0; H1-1; H1-12; H1-2; H1A; H1B; H1c; H1F0; H1f1; H1F2; H1F3; H1F5; H1FV; H1s-1; H1s-3; H1var1; H1var3; Hils1; His1a; HIST1; HIST1H1A; HIST1H1B; HIST1H1B antibody; HIST1H1C; HIST1H1D; HIST1H1E; Histone 1 antibody; histone 1, family 5; histone 1, H1a; histone 1, H1b; histone 1, H1c; histone cluster 1, H1a; histone cluster 1, H1b; histone cluster 1, H1c; histone cluster 1, H1e; histone H1; histone H1'; Histone H1(0); histone H1.0; Histone H1.0, N-terminally processed; Histone H1.1; histone H1.2; Histone H1.3; Histone H1.5; histone H1a; Histone H1b; histone H1c; Histone H1d; histone H1-like protein in spermatids 1; histone H1s-1; Histone H1s-2; Histone H1s-3; histone linker H1 domain, spermatid-specific 1 (pseudogene); spermatid-specific linker histone H1-like protein; TISP64 |
| Gensymboler | H1-0, H1-1, H1-2, H1-5 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?