missing translation for 'onlineSavingsMsg'
Learn More

Phospho-MARCKS (Ser93, Ser97, Ser104) Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10018-3-AP-100UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16876163

  • 3350.00 SEK / 100µL
Estimated Shipment: 23-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is raised against phospho-Marcks with S93, S97 and S104 phosphorylated.

MARCKS is a substrate for protein kinase C. It is localized to the plasma membrane and is an actin filament crosslinking protein. Phosphorylation by protein kinase C or binding to calcium-calmodulin inhibits its association with actin and with the plasma membrane, leading to its presence in the cytoplasm. The protein is thought to be involved in cell motility, phagocytosis, membrane trafficking and mitogenesis.
Specifications

Specifications

Phospho-MARCKS (Ser93, Ser97, Ser104)
Polyclonal
Unconjugated
MARCKS
Macs, Marcks, phospho-Marcks, PKCSL
Rabbit
Antigen Affinity Chromatography
RUO
17118, 4082
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P26645, P29966
Marcks
Peptide
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers