missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PDX1 (Ser66) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564702
Additional Details : Weight : 0.01000kg
Description
Phospho-PDX1 (Ser66) Polyclonal Antibody detects endogenous levels of PDX1 only when phosphorylated at Ser66.
Phospho-PDX1 (Ser66) Polyclonal antibody specifically detects Phospho-PDX1 (Ser66) in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Phospho-PDX1 (Ser66) | |
Polyclonal | |
Unconjugated | |
PDX1 | |
DLDBP; E3BP; Glucose-sensitive factor; GSF; Idx1; IDX-1; Insulin promoter factor 1; insulin promoter factor 1, homeodomain transcription factor; insulin promotor factor 1; insulin upstream factor 1; Ipf1; IPF-1; Islet/duodenum homeobox 1; islet/duodenum homeobox-1; IUF1; IUF-1; MODY4; PAGEN1; Pancreas/duodenum homeobox protein 1; pancreatic and duodenal homeobox 1; pancreatic and duodenal homeobox gene 1; pancreatic-duodenal homeobox factor 1; PDHX; PDX1; pdx-1; somatostatin transactivating factor 1; somatostatin transcription factor 1; somatostatin-transactivating factor 1; Stf1; STF-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
18609, 29535, 3651 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P52945, P52946, P52947 | |
PDX1 | |
A synthesized peptide derived from human PDX1 (Phospho-Ser66) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |