Läs mer
Phospho-PYK2/FAK2 (Y402) Mouse anti-Human, Clone: 592918, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB6210-SP
Encompass
Produktkod. 15737904
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
PYK2/FAK2 Monoclonal specifically detects PYK2/FAK2 in Human samples. It is validated for Western Blot, Simple Western.
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer