missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Phospho-RAC1/CDC42 (Ser71) Recombinant Rabbit Monoclonal Antibody (20H14L7)

Produktkod. 15612035
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
Förpackningsstorlek:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15612035 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15612035 Leverantör Invitrogen™ Leverantörsnummer 701942

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Pig, Rat, Monkey and Mouse. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Rac1 is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for the Rac1 gene.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Phospho-RAC1/CDC42 (Ser71)
Användningsområden Western Blot, Immunocytochemistry
Klassificering Recombinant Monoclonal
Klona 20H14L7
Koncentration 0.5 mg/mL
Konjugera Unconjugated
Formulering PBS with 0.09% sodium azide; pH 7.2
Gen CDC42
Genaccessionsnr. P60953, P63000
Gene Alias AI747189; AL023026; AU018915; CDC42; CDC42Hs; Cell division control protein 42 homolog; cell division cycle 42; cell division cycle 42 (GTP binding protein); cell division cycle 42 homolog; cell migration-inducing gene 5 protein; D5Ertd559e; dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein, 25kD)); dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein, 25kD)); G25K; G25K GTP-binding protein; growth-regulating protein; GTP binding protein, 25kDa; MIG5; p21-Rac1; Rac1; Rac-1; Ras-like protein TC25; RAS-related C3 botulinum substrate 1; ras-related C3 botulinum toxin substrate 1; ras-related C3 botulinum toxin substrate 1 (rho family small GTP binding protein Rac1); ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1); rho family, small GTP binding protein Rac1; small GTP binding protein CDC42; TC25; TC-25; TKS
Gensymboler CDC42, RAC1
Värd art Rabbit
Immunogen Peptide corresponding to Human CDC42 (aa 68-76).
Reningsmetod Protein A
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 5879, 998
Målarter Human
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.