missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Phospho-RyR2 (Ser2814) Polyclonal Antibody
GREENER_CHOICE

Produktkod. 16691005
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16691005 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16691005 Leverantör Invitrogen™ Leverantörsnummer PA5104558

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of RYR2 only when phosphorylated at Ser2814.

Dihydropyridine receptor (DHPR) is a surface membrane protein critical for the excitation-contraction coupling of striated muscle. DHPR and the sarcoplasmic reticulum ryanodine receptor (RyR) are two key components of the intracellular junctions, where depolarization of the surface membrane is converted into the release of Ca2+ from internal stores. The alpha1-subunit of the DHPR contains a cytoplasmic loop which is thought to be involved in the interactions with RyR. Phosphorylation of the DHPR alpha1-subunit is also thought to play a role in the functional interaction of DHPR and RyR. Mutation in DHPR alpha1 results in excitation-contraction uncoupling, leading to muscular dysgenesis, a complete inactivity in developing skeletal muscles. Cells that do not express RyR also lack excitation-contraction coupling and exhibit a severalfold reduction in Ca2+ current density.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Phospho-RyR2 (Ser2814)
Användningsområden Immunohistochemistry (Paraffin), Western Blot
Klassificering Polyclonal
Koncentration 1 mg/mL
Konjugera Unconjugated
Formulering PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gen Ryr2
Genaccessionsnr. Q92736
Gene Alias 9330127I20Rik; ARVC2; ARVD2; calcium release channel; cardiac muscle ryanodine receptor; Cardiac muscle ryanodine receptor-calcium release channel; cardiac ryanodine receptor 2; cardiac-type ryanodine receptor; hRYR-2; islet-type ryanodine receptor; kidney-type ryanodine receptor; Ryanodine receptor; ryanodine receptor 2; ryanodine receptor 2 (cardiac); ryanodine receptor 2, cardiac; ryanodine receptor type 2; ryanodine receptor type II; RyR; RYR2; RYR-2; Type 2 ryanodine receptor; VTSIP
Gensymboler Ryr2
Värd art Rabbit
Immunogen A synthesized peptide derived from human RYR2(Accession Q92736), corresponding to amino acid residues around phosphorylated Ser2814.
Reningsmetod Sequential chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 403615, 6262
Målarter Human, Canine
Innehåll och lagring -20°C
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.