Learn More
Phospho-SMAD3 (Ser423, Ser425) Recombinant Rabbit Monoclonal Antibody (16H5L12), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen 702292
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with Monkey, Sheep, Rat and Pig Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Phospho-SMAD3 (Ser423, Ser425) Recombinant Monoclonal antibody specifically detects Phospho-SMAD3 (Ser423, Ser425) in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Phospho-SMAD3 (Ser423, Ser425) | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P84022 | |
SMAD3 | |
Peptide corresponding to Human SMAD3 (aa 418-425) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
16H5L12 | |
Unconjugated | |
SMAD3 | |
AU022421; DKFZp586N0721; DKFZp686J10186; hMAD-3; hSMAD3; HSPC193; HsT17436; JV15-2; LDS1C; LDS3; MAD (mothers against decapentaplegic, Drosophila) homolog 3; MAD homolog 3; MAD homolog 3a; mad homolog JV15-2; mad protein homolog; MAD, mothers against decapentaplegic homolog 3; mad3; MAD-3; madh3; madh3a; madh3-A; MGC60396; mMad3; mothers against decapentaplegic homolog 3; mothers against decapentaplegic homolog 3a; mothers against DPP homolog 3; SMA- and MAD-related protein 3; SMAD; Smad 3; SMAD family member 3; SMAD family member 3 L homeolog; SMAD family member 3a; SMAD, mothers against DPP homolog 3; Smad3; smad3.L; smad3a; TGF beta response effector Smad3; TGF-beta response effector Smad3; transcription factor; transcription factor Smad3; wu:fa99e03; XELAEV_18018454mg; XenMLP; Xmad3; XSmad3 | |
Rabbit | |
Protein A | |
RUO | |
4088 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |