missing translation for 'onlineSavingsMsg'
Learn More

Phospho-STAT3 (Tyr705) Mouse anti-Human, Mouse, eFluor® 450, Clone: LUVNKLA, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-9033-42

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15559826

  • 2945.00 SEK / 100 tests


Estimated Shipment
04-01-2021
Add to basket

Description

Description

Applications Reported: This LUVNKLA antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This LUVNKLA antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Staining Protocol: We recommend using Protocol C: Two-step protocol: Fixation/Methanol. Protocol A: Two-step protocol: intracellular (cytoplasmic) proteins and Protocol B: One-step protocol: intracellular (nuclear) proteins cannot be used. All Protocols can be found in the Flow Cytometry Protocols: "Staining Intracellular Antigens for Flow Cytometry Protocol" located in the Best Protocols Section under the Resources tab online. eFluor™ 450 is an alternative to Pacific Blue™. eFluor™ 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered. STAT3 belongs to the family of STAT (signal transducers and activators of transcription) proteins which are phosphorylated by receptor associated kinases, translocate to the nucleus, and act as transcription factors in response to cytokines and growth factors. Coactivators such as CREB-binding protein are required for the transcriptional activation by STAT3. STAT3 can also be activated by Interferon-alpha, Interferon-gamma, EGF, PDGF and IL6. Phosphorylation on tyrosine 705 by JAK1 and JAK2 is essential for STAT3 dimer formation, nuclear translocation, and DNA binding activity. In humans, the STAT3 gene is located on the q arm of chromosome 17. STAT3 has been shown to be activated by IFN-alpha but not IFN-beta. The transcription factors associated with STAT3 are c-Jun and cyclic AMP-responsive enhancer binding protein (CREB). STAT3 mediates the expression of a variety of genes in response to cell stimuli, and thus plays a key role in many cellular processes such as cell growth and apoptosis. The small GTPase Rac1 has been shown to bind and regulate the activity of STAT3 while the PIAS3 protein is a specific inhibitor of STAT3. Three alternatively spliced transcript variants encoding distinct isoforms of STAT3 have been described. Deletion of the STAT3 gene in knock-out mice was lethal at the early embryonic stage.
Specifications

Specifications

Phospho-STAT3 (Tyr705)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P42227, P40763
Signal Transducers and Activators of Transcription 3, Acute-phase response factor
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Flow Cytometry
LUVNKLA
eFluor™ 450
STAT3
Liquid
Stat3
IgG2b, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
20848, 6774
Videos
SDS
Documents

For Research Use Only.