Learn More
Phospho-STAT3 (Tyr705) Monoclonal Antibody (LUVNKLA), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1294.00 SEK - 3640.00 SEK
Specifications
Antigen | Phospho-STAT3 (Tyr705) |
---|---|
Clone | LUVNKLA |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15559826
|
Affymetrix eBioscience
48-9033-42 |
100 Tests |
3640.00 SEK
100 tests |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15549826
|
Affymetrix eBioscience
48-9033-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This LUVNKLA monoclonal antibody recognizes human and mouse signal transducer and activator of transcription 3 (STAT3) when phosphorylated on tyrosine 705 (Y705). The STAT family represents seven transcription factors (STATs 1, 2, 3, 4, 5A, 5B, and 6) that are involved in many cellular processes including apoptosis, cell differentiation, and proliferation in a cell type- and cytokine-specific manner. STAT proteins are activated by ligand binding to cytokine receptors that associate with Janus kinase (JAK) family members. Following their phosphorylation by JAKs, STAT proteins translocate to the nucleus where they bind to DNA and regulate transcription of specific genes in a cell type- and cytokine-specific manner. STAT3 is activated downstream of numerous cytokines including interferons, IL-5, IL-6, IL-10, and LIF. STAT3 is important for the differentiation of Th17 cells and mediates a variety of cellular processes including cell growth and survival. The importance of STAT3 is highlighted by both loss-of-function and gain-of-function mutations. Deletion of STAT3 in T cells results in decreased IL-6- and IL-2-mediated proliferation, while deletion of STAT3 in neutrophils and macrophages results in increased susceptibility to LPS-induced endotoxic shock and increased production of the pro-inflammatory cytokines IL-6 and TNF alpha. Hyper STAT3 activity is associated with poor prognosis of many different cancers.
Phospho-STAT3 (Tyr705) Monoclonal antibody specifically detects Phospho-STAT3 (Tyr705) in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
Phospho-STAT3 (Tyr705) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P40763, P42227 | |
STAT3 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
STAT3 |
LUVNKLA | |
Flow Cytometry | |
eFluor 450 | |
Mouse | |
Human, Mouse | |
20848, 6774 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.