missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Phospho-VAV2 (Tyr172) Polyclonal Antibody
GREENER_CHOICE

Produktkod. 17856858
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17856858 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17856858 Leverantör Invitrogen™ Leverantörsnummer PA5143788

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

This antibody was cross-adsorbed to unphosphorylated Vav2 (Tyr-172) peptide before affinity purification using phospho-Vav2 (Tyr-172) peptide (without carrier). The antibody detects a 100 kDa* band corresponding to Vav on SDS-PAGE immunoblots of human A431 cells treated with EGF and detects Vav1, Vav2, and Vav3 in Jurkat cells treated with pervanadate. The antibody does not detect these Vavs after treatment with alkaline phosphatase.

VAV2 is the second member of the VAV guanine nucleotide exchange factor family of oncogenes. Unlike VAV1, which is expressed exclusively in hematopoietic cells, VAV2 transcripts were found in most tissues. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Phospho-VAV2 (Tyr172)
Användningsområden Western Blot, Immunocytochemistry
Klassificering Polyclonal
Koncentration 0.125 mg/mL
Konjugera Unconjugated
Formulering PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gen Vav2
Genaccessionsnr. P52735, Q60992
Gene Alias 2810040F13Rik; AI847175; Guanine nucleotide exchange factor VAV2; Oncogene VAV2; vav 2 guanine nucleotide exchange factor; vav 2 oncogene; vav guanine nucleotide exchange factor 2; VAV2; VAV-2; Vav2 oncogene
Gensymboler Vav2
Värd art Rabbit
Immunogen Phospho-Vav2 (Tyr-172) synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding Tyr-172 in mouse Vav2. This sequence has high homology with similar regions in rat and human Vav2, and has significant homology to the conserved site in Vav1 and Vav3.
Reningsmetod Antigen affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 22325, 296603, 7410
Målarter Human, Mouse, Rat
Innehåll och lagring -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.