Läs mer
Phosphoglycolic Acid, TRC
Produktkod. 30413845
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30413845 25 mg
30425310 5 mg
30424631 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30413845 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP35820025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13147-57-4
Molekylformel C2H5O6P
Molekylvikt (g/mol) 156.03
Synonym 2-(Phosphonooxy)acetic Acid, (Phosphonooxy)acetic Acid, Glycolic Acid Phosphate, Glycolic Acid, di-H Phosphate, Glycolic Acid, Dihydrogen Phosphate, 2-Phosphoglycolic Acid, Glycophosphoric Acid
IUPAC-namn 2-phosphonooxyacetic acid
LEDER OC(=O)COP(=O)(O)O
Specifikationer
Kemiskt namn eller material Phosphoglycolic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C2H5O6P/c3-2(4)1-8-9(5,6)7/h1H2,(H,3,4)(H2,5,6,7)
Formel vikt 155.9824
Procent renhet >95%