Läs mer
Phosphonic Acid, 4-Pyridinyl-, Dimethyl Ester, TRC
Produktkod. 30423565
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30423565 100 mg
1 options
Produktkod. 30423565 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP228138100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phosphonic Acid','4-Pyridinyl-','Dimethyl Ester
CAS 78133-46-7
Molekylformel C7H10NO3P
Kvantitet 100 mg
Synonym dimethyl pyridin-4-ylphosphonate
InChI formel InChI=1S/C7H10NO3P/c1-10-12(9,11-2)7-3-5-8-6-4-7/h3-6H,1-2H3
LEDER O=P(OC)(OC)C=1C=CN=CC1
IUPAC-namn dimethyl pyridin-4-ylphosphonate
Molekylvikt (g/mol) 187.13
Formel vikt 187.04