Läs mer
Phosphoric Acid Diethyl Octyl Ester, TRC
Produktkod. 30419179
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30419179 500 mg
30423000 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30419179 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP359480500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 20195-13-5
Molekylformel C12H27O4P
Molekylvikt (g/mol) 266.32
Synonym Ethyl Octyl Phosphate ((EtO)2(C8H17O)PO) (6CI,7CI), O,O-Diethyl O-octyl Phosphate, Octyl Diethyl Phosphate
IUPAC-namn diethyl octyl phosphate
LEDER CCCCCCCCOP(=O)(OCC)OCC
Specifikationer
Kemiskt namn eller material Phosphoric Acid Diethyl Octyl Ester
Kvantitet 500 mg
InChI formel InChI=1S/C12H27O4P/c1-4-7-8-9-10-11-12-16-17(13,14-5-2)15-6-3/h4-12H2,1-3H3
Formel vikt 266.1647