Learn More
Phosphoric Acid, Diluted, 10% Solution, Multi-Compendial, N.F., J.T.Baker™
Brand:  JT Baker 5683-08
Additional Details : Weight : 19.00000kg Transport : UN number : 1805 Chem class : 8 Pack group : III
Quantity 19L
Packaging Poly Drum
 View more versions of this product
Product Code. 15518364
  • 6802.43 SEK / 19 L
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Specifications
Chemical Identifiers
Phosphoric Acid
Diluted, 10% Solution
H3PO4
7664-38-2
Multi-Compendial/NF
98
Specifications
Phosphoric Acid
Diluted, 10% Solution
Multi-Compendial/NF
98
1L = 1.05kg
19L
7664-38-2
H3PO4
Poly Drum
Videos