missing translation for 'onlineSavingsMsg'
Läs mer

Phosphoric Acid Dioctyl Ester Potassium Salt, TRC

Produktkod. 30424633
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424633 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Product Code. 30424633 Supplier Toronto Research Chemicals Supplier No. TRCP359420500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Phosphoric Acid Dioctyl Ester Potassium Salt
CAS 19045-78-4
Molekylformel C16H34KO4P
Kvantitet 500 mg
Synonym Dioctyl Phosphate Potassium Salt, Potassium Dioctyl Phosphate
InChI formel InChI=1S/C16H35O4P.K/c1-3-5-7-9-11-13-15-19-21(17,18)20-16-14-12-10-8-6-4-2;/h3-16H2,1-2H3,(H,17,18);/q;+1/p-1
LEDER O=P(OCCCCCCCC)(OCCCCCCCC)[O-].[K+]
IUPAC-namn potassium dioctyl phosphate
Molekylvikt (g/mol) 360.51
Formel vikt 360.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.