Läs mer
Phosphoric Acid Dioctyl Ester Potassium Salt, TRC
Produktkod. 30419176
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30419176 1 g
1 options
Produktkod. 30419176 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3594201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phosphoric Acid Dioctyl Ester Potassium Salt
CAS 19045-78-4
Molekylformel C16H34KO4P
Kvantitet 1 g
Synonym Dioctyl Phosphate Potassium Salt, Potassium Dioctyl Phosphate
InChI formel InChI=1S/C16H35O4P.K/c1-3-5-7-9-11-13-15-19-21(17,18)20-16-14-12-10-8-6-4-2;/h3-16H2,1-2H3,(H,17,18);/q;+1/p-1
LEDER O=P(OCCCCCCCC)(OCCCCCCCC)[O-].[K+]
IUPAC-namn potassium dioctyl phosphate
Molekylvikt (g/mol) 360.51
Formel vikt 360.18