missing translation for 'onlineSavingsMsg'
Läs mer

Phosphoric Acid Tributyl Ester-d27, TRC

Product Code. 30425533
Change view
Click to view available options
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30425533 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30425533 Supplier Toronto Research Chemicals Supplier No. TRCP359447100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Tributyl phosphate-d27
CAS 61196-26-7
Anteckningar om renhetsgrad GC
Molekylformel C12 D27 O4 P
Kvantitet 100 mg
Synonym 1-Butan-1,1,2,2,3,3,4,4,4-d9-ol','phosphate (3:1) (9CI), Tributyl Phosphate D27
InChI formel InChI=1S/C12H27O4P/c1-4-7-10-14-17(13,15-11-8-5-2)16-12-9-6-3/h4-12H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2
LEDER [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])OP(=O)(OC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])OC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
IUPAC-namn tris(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl) phosphate
Molekylvikt (g/mol) 293.48
Formel vikt 293.334
Procent renhet >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.