Learn More
Phosphorus (P) Standard Solution, 10000 μg/mL in 5% HNO3, VHG
Product Code. 30003116
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. Quantity Unit Size
30003116 100 mL 100mL
1 options
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 30003116 Supplier VHG Labs Supplier No. VHGTPN100
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Specifications
CAS 7723-14-0
Analyte or Component Names Phosphorus
Shipping Condition Room Temperature
Concentration 10000 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/P
Solution Type Single Solution
SMILES [P]
IUPAC Name Phosphorus
Solvent or Matrix Dilute HNO3
Industry Type Petrochemical Testing
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Phosphorus 10000 μg/mL