Learn More
Hanna Instruments™ Phosphorus Reagent Sets, For Colorimetric Water Testing
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Phosphorus Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
Brand:  Hanna Instruments™ HI93758A50
Product Code. 11632690
  • 2430.00 SEK / Each
Estimated Shipment: 10-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
These high quality reagents are designed to follow accepted test methods for colorimetric determination of various forms of phosphorus. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
  • Reagent sets available for determinations of phosphorus, total phosphorus (high or low range), acid hydrolyzable phosphorus and reactive phosphorus (low or high range)
  • All sets except basic Phosphorus Reagents (HI93706-01) contain 50 ready-to-use vials as well reagent packets/dropper bottles
  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability
Specifications
Specifications
Reactive phosphorus determination
Ready-to-use vials of HI93758A-0 and individual packets of HI93758-0
1 Set
EPA method 365.2 and Standard Methods, 20th edition, 4500-P E - ascorbic acid method
Compatible Hanna benchtop and portable photometers
50
0.00 to 5.00mg/L (ppm) phosphorus
Water Testing Kit