missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus Standard For ICP, 10,000μg/mL in H2O, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

943.00 SEK - 2070.00 SEK

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name azanium;dihydrogen phosphate
SMILES [NH4+].OP(O)([O-])=O
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10172532
View Documents Promotion Details
SPEX Certiprep
PLP9-3Y
125mL Triple-leached LDPE bottle
943.00 SEK
125mL
View alternative products
Save Up to 
»
Estimated Shipment: 05-07-2022
to see stock.
Please to purchase this item. Need a web account? Register with us today!
10467112
View Documents Promotion Details
SPEX Certiprep
PLP9-3X
500mL Triple-leached LDPE bottle
2070.00 SEK
500mL
View alternative products
Save Up to 
»
Estimated Shipment: 05-07-2022
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
P10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

7722-76-1
115.03
LFVGISIMTYGQHF-UHFFFAOYSA-N
CHEBI:62982
[NH4+].OP(O)([O-])=O
H6NO4P
MFCD00003396
24402
azanium;dihydrogen phosphate

Specifications

Phosphorus Standard
H6NO4P
LFVGISIMTYGQHF-UHFFFAOYSA-N
azanium;dihydrogen phosphate
24402
Assurance
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
125mL
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
7722-76-1,7732-18-5
MFCD00003396
[NH4+].OP(O)([O-])=O
115.03
CHEBI:62982
Triple-leached LDPE bottle
H2O
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
SDS
Product Certifications

Product intended for laboratory use only