Learn More
Phosphorus(V) oxychloride, 99%, Thermo Scientific Chemicals
Catalog Number 15490987
Click to view available options
Quantity:
250 g
1 kg
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
10025-87-3
Cl3OP
153.323
MFCD00011443
XHXFXVLFKHQFAL-UHFFFAOYSA-N
phosphorus oxychloride, phosphoryl chloride, phosphoric trichloride, phosphoryl trichloride, phosphoroxychloride, trichlorophosphine oxide, phosphorus oxytrichloride, fosforoxychlorid, phosphorous oxychloride, phosphorylchlorid
24813
CHEBI:30336
O=P(Cl)(Cl)Cl
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
10025-87-3
Cl3OP
153.323
MFCD00011443
XHXFXVLFKHQFAL-UHFFFAOYSA-N
phosphorus oxychloride, phosphoryl chloride, phosphoric trichloride, phosphoryl trichloride, phosphoroxychloride, trichlorophosphine oxide, phosphorus oxytrichloride, fosforoxychlorid, phosphorous oxychloride, phosphorylchlorid
24813
CHEBI:30336
O=P(Cl)(Cl)Cl
Phosphorus(V) oxychloride is widely used as a dehydrating agent and a chlorinating agent. Useful for the conversion of amide to nitriles, Bischler-Napieralski reaction, Vilsmeier Haak reaction, and other reactions producing commercially important intermediates. Acts a source of liquid phosphorus in semiconductor industry. Manufacture of trialkyl phosphate esters and triarylphosphate esters, including commercially popular tricresyl phosphate,which is useful as a plasticizer, flame retardant, gasoline additive, hydrolytic fluid, etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Boiling Point 105.8°C
Melting Point 1.25°C
Color Colorless
Physical Form Liquid
Quantity 1 kg
Assay Percent Range 99%
Linear Formula POCl3
UN Number UN1810
Merck Index 14,7349
Solubility Information Phosphorus(V) oxychloride is soluble in many organic solvents.
Formula Weight 153.33
Percent Purity 99%
Odor Pungent
Refractive Index 1.46
Sensitivity Moisture sensitive
Appearance Colorless liquid
Vapor Pressure 36 hPa (27mm Hg) at 20°C
Density 1.685 g/cm3 at 20°C
Chemical Name or Material Phosphorus(V) oxychloride