missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Phosphotyrosine Monoclonal Antibody (P-Tyr-01)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA110443
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This antibody detects tyrosine phosphorylation in activated cells.
The role of tyrosine phosphorylation in transduction of the mitogenic signal from transmembrane receptors and in transformation by oncogene tyrosine kinases has been the subject of intense investigation for several years. While the phosphorylation of specific tyrosine residues has been shown to be a primary mechanism of signal transduction during normal mitogenesis, cell cycle progression and oncogenic transformation, its role in other areas such as differentiation and gap junction communication, is a matter of active and ongoing research. Antibodies that specifically recognize phosphorylated tyrosine residues have proved to be invaluable to the study of tyrosine -phosphorylated proteins and the biochemical pathways in which they function. The fluorescein (FITC) conjugate of clone PY20 anti-phosphotyrosine is especially useful for the detection of these P-Tyr proteins in immunohistochemical and immunocytochemical protocols in situations wherein the use of a secondary antibody would complicate detection of the protein(s) of interest.
Specifikationer
| Phosphotyrosine | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| Mouse | |
| Protein A | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| P-Tyr-01 | |
| Unconjugated | |
| Phosphotyrosine; pTyr; pY | |
| Phosphotyrosine conjugated to bovine serum albumin. | |
| 100 μg | |
| Primary | |
| 4°C, do not freeze | |
| Liquid | |
| Chemical |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering