missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Photoluminescent Warning Striped Tape - Left
Brand: Brady™ 263367
Additional Details : Weight : 0.14000kg
Specifications
Warning Tape: Standard Luminous Intensity Red Photoluminescent. Striped - Right | |
Photoluminescent Warning Striped Tape | |
12 m | |
0.01 kg |
Red | |
1 Ea. | |
30 mm |