Läs mer
Palintest™ Photometer Kit
Produkttyp:
Fotometer 7100-sats
Fotometer 7500-kit
Förpackningsstorlek:
Varje
Beskrivning
- Includes: 7100 Model Standard, with accessories
- Photometer kit for 7100 device
- Comes complete in case with six glass tubes and one dilution tube
- Power supply
- 3 AA batteries
- L x W x H: 146 x 275 x 75 mm
- Weight: 975 g
Specifikationer
Specifikationer
| Inkluderar | 7500 Model with accessories |
| Produkttyp | Fotometer 7500-kit |