Läs mer
Phoxim, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Phoxim |
| CAS | 14816-18-3 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C12 H15 N2 O3 P S |
| Kvantitet | 1 g |
| Synonym | Phosphorothioic acid','O-[(cyanophenylmethylene)azanyl] O,O-diethyl ester (ACI), 3,5-Dioxa-6-aza-4-phosphaoct-6-ene-8-nitrile','4-ethoxy-7-phenyl-','4-sulfide (9CI), Glyoxylonitrile','phenyl-','oxime O,O-diethyl phosphorothioate (8CI), Phosphorothioic acid','O,O-diethyl ester','O-[(α-cyanobenzylidene)amino] deriv. (8CI), B 77488, Basileum SI, Basileum SI 84, Basileum SI 84EC, BAY 5621, BAY 77488, Bayer 5621, Bayer 77488, Baythion, Benzoyl cyanide O-(diethoxyphosphinothioyl)oxime, Folaton, Jiubaojing, O,O-Diethyl O-(α-cyanobenzylideneamino) phosphorothioate, Phenylglyoxylonitrile oxime O,O-diethyl phosphorothioate, Phoxim, Sebacil, Tomiguard PE 50, Toyothion, Valexon, Volaton, Volaton bjdse 20, Xylasan VL |
| InChI formel | InChI=1S/C12H15N2O3PS/c1-3-15-18(19,16-4-2)17-14-12(10-13)11-8-6-5-7-9-11/h5-9H,3-4H2,1-2H3 |
| LEDER | CCOP(=S)(OCC)ON=C(C#N)c1ccccc1 |
| IUPAC-namn | N-diethoxyphosphinothioyloxybenzenecarboximidoyl cyanide |
| Molekylvikt (g/mol) | 298.3 |
| Visa mer |